Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007361.3 | NP_031387.3 | 4 | 1215 | |
XM_005267405.4 | XP_005267462.1 | 4 | 1218 | ||
XM_005267406.4 | XP_005267463.1 | 4 | 1218 | ||
XM_005267407.4 | XP_005267464.1 | 4 | 1218 | ||
GenBank mRNA | AJ223500.1 | - | 4 | 981 | |
AK291652.1 | - | 4 | 1043 | ||
AK300462.1 | - | 5 | 853 | ||
AL701794.1 | - | 3 | 368 | ||
BX648241.1 | - | 5 | 988 | ||
D86425.1 | - | 4 | 984 |