Gene Transcripts
- Gene Symbol
- PSME4
- Entrez Gene ID
- 23198
- Gene Name
- proteasome activator subunit 4
- Gene Aliases
- PA200
- Chromosome Location
- Chr. 2: 53864067 - 53970840 on Build GRCh38
- UniGene ID
- Hs.413801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014614.2 |
NP_055429.2
|
34, 35 |
3976 |
|
XM_006711969.1 |
XP_006712032.1
|
33, 34 |
3553 |
|
XM_011532705.2 |
XP_011531007.1
|
35, 36 |
4000 |
|
XM_011532706.2 |
XP_011531008.1
|
35, 36 |
4000 |
|
XM_011532707.2 |
XP_011531009.1
|
34, 35 |
3934 |
|
GenBank mRNA |
AK025517.1 |
- |
3, 4 |
320 |
|
AK026085.1 |
- |
8, 9 |
973 |
|
AY894754.1 |
- |
33, 34 |
3825 |
|
AY894755.1 |
- |
15, 16 |
1664 |
|
BC112169.1 |
- |
33, 34 |
3612 |
8327637 |
BC113668.1 |
- |
33, 34 |
3612 |
8322720 |
BC143739.1 |
- |
33, 34 |
3612 |
|
D38521.1 |
- |
34, 35 |
3786 |
|
View Full Product Details