Gene Transcripts

Gene Symbol
POP1
Entrez Gene ID
10940
Gene Name
POP1 homolog, ribonuclease P/MRP subunit
Gene Aliases
Chromosome Location
Chr. 8: 98117293 - 98159841 on Build GRCh38
UniGene ID
Hs.252828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145860.1 NP_001139332.1 5 732
NM_001145861.1 NP_001139333.1 5 672
NM_015029.2 NP_055844.2 5 660
XM_011516801.2 XP_011515103.1 5 732
GenBank mRNA AK291434.1 - 5 729
BC011529.1 - 5 660 3853321
D31765.1 - 2 271

View Full Product Details