Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 22 | 2738 | |
NM_003664.4 | NP_003655.3 | 22 | 2726 | ||
XM_005248618.3 | XP_005248675.1 | 22 | 2698 | ||
XM_005248619.4 | XP_005248676.1 | 22 | 2698 | ||
XM_017010001.1 | XP_016865490.1 | 22 | 2478 | ||
GenBank mRNA | AK225908.1 | - | 22 | 2602 | |
AK291201.1 | - | 22 | 2667 | ||
BC038444.1 | - | 22 | 2687 | 3914400 | |
BX538041.1 | - | 22 | 2676 | ||
KU178426.1 | - | 22 | 2550 | ||
U81504.1 | - | 22 | 2642 | ||
U91931.1 | - | 22 | 2600 |