Gene Transcripts

Gene Symbol
CCR9
Entrez Gene ID
10803
Gene Name
C-C motif chemokine receptor 9
Gene Aliases
CC-CKR-9, CDw199, GPR-9-6, GPR28
Chromosome Location
Chr. 3: 45886504 - 45903177 on Build GRCh38
UniGene ID
Hs.225946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256369.1 NP_001243298.1 4 2381
NM_006641.3 NP_006632.2 2 2246
NM_031200.2 NP_112477.1 3 2295
XM_011533312.2 XP_011531614.1 3 3008
GenBank mRNA AF145439.1 - 3 2272
AF145440.1 - 2 2191
BC069678.1 - 3 2256 7262627

View Full Product Details