Gene Transcripts

Gene Symbol
NOL4
Entrez Gene ID
8715
Gene Name
nucleolar protein 4
Gene Aliases
CT125, HRIHFB2255, NOLP
Chromosome Location
Chr. 18: 33851100 - 34224834 on Build GRCh38
UniGene ID
Hs.514795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198546.1 NP_001185475.1 9 1769
NM_001198547.1 NP_001185476.1 10 1850
NM_001198548.1 NP_001185477.1 8 1655
NM_001198549.1 NP_001185478.1 6 1065
NM_001282527.1 NP_001269456.1 10 1699
NM_003787.4 NP_003778.2 10 1961
XM_006722563.2 XP_006722626.1 11 2718
XM_006722564.2 XP_006722627.1 9 1511
XM_006722565.1 XP_006722628.1 11 1737
XM_011526237.1 XP_011524539.1 11 2715
XM_011526238.1 XP_011524540.1 10 2327
XM_011526239.2 XP_011524541.1 9 1719
XM_017026052.1 XP_016881541.1 9 1549
XM_017026053.1 XP_016881542.1 10 1701
XM_017026054.1 XP_016881543.1 11 1829
XM_017026055.1 XP_016881544.1 10 2176
XM_017026056.1 XP_016881545.1 10 1430
XM_017026057.1 XP_016881546.1 11 2293
XM_017026058.1 XP_016881547.1 10 1440
XM_017026059.1 XP_016881548.1 11 3308
XM_017026060.1 XP_016881549.1 5 887
GenBank mRNA AB015339.1 - 5 658
AB017800.1 - 10 1971
AK091463.1 - 10 1548
AK296539.1 - 6 1048
AK297185.1 - 9 1423
AK299850.1 - 10 1472
AK312562.1 - 10 1296
BC000313.2 - 8 1341 2821116
BT006763.1 - 7 1015
CR456730.1 - 7 1015

View Full Product Details