Gene Transcripts
- Gene Symbol
- PEX11B
- Entrez Gene ID
- 8799
- Gene Name
- peroxisomal biogenesis factor 11 beta
- Gene Aliases
- PEX11-BETA, PEX14B
- Chromosome Location
- Chr. 1: 145911348 - 145918924 on Build GRCh38
- UniGene ID
- Hs.714608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184795.1 |
NP_001171724.1
|
2, 3 |
221 |
|
NM_003846.2 |
NP_003837.1
|
2, 3 |
415 |
|
NR_073491.1 |
|
2, 3 |
411 |
|
NR_073492.1 |
|
2, 3 |
405 |
|
NR_073493.2 |
|
2, 3 |
627 |
|
GenBank mRNA |
AB018080.1 |
- |
2, 3 |
197 |
|
AF093670.1 |
- |
2, 3 |
195 |
|
AK023991.1 |
- |
2, 3 |
179 |
|
AK301983.1 |
- |
2, 3 |
181 |
|
BC011963.1 |
- |
2, 3 |
195 |
3866690 |
BG423552.1 |
- |
|
155 |
|
CB155824.1 |
- |
2, 3 |
221 |
|
CR542047.1 |
- |
2, 3 |
179 |
|
DB054933.1 |
- |
2, 3 |
415 |
|
HY108328.1 |
- |
2, 3 |
198 |
|
View Full Product Details