Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039711.1 | NP_001034800.1 | 2 | 381 | |
NM_001039712.1 | NP_001034801.1 | 3 | 352 | ||
NM_032998.2 | NP_127491.1 | 3 | 414 | ||
XM_005245597.3 | XP_005245654.1 | 3 | 325 | ||
XM_005245598.4 | XP_005245655.1 | 3 | 1464 | ||
XM_005245599.2 | XP_005245656.1 | 3 | 368 | ||
XM_005245600.2 | XP_005245657.1 | 2 | 476 | ||
XM_017002802.1 | XP_016858291.1 | 3 | 1464 | ||
GenBank mRNA | AF043733.1 | - | 1 | 229 | |
AF064605.3 | - | 2 | 266 | ||
AF083236.1 | - | 1 | 179 | ||
AF100341.1 | - | 1 | 179 | ||
AJ010973.1 | - | 1 | 179 | ||
AK001497.1 | - | 3 | 317 | ||
AK022531.1 | - | 3 | 302 | ||
BC013910.2 | - | 3 | 394 | 3504847 | |
BC016724.1 | - | 3 | 315 | 4077893 | |
BG565440.1 | - | 3 | 353 | ||
CR536556.1 | - | 1 | 179 |