Gene Transcripts
- Gene Symbol
- TRIM10
- Entrez Gene ID
- 10107
- Gene Name
- tripartite motif containing 10
- Gene Aliases
- HERF1, RFB30, RNF9
- Chromosome Location
- Chr. 6: 30151939 - 30163216 on Build GRCh38
- UniGene ID
- Hs.709483
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006778.3 |
NP_006769.2
|
4, 5 |
851 |
|
NM_052828.2 |
NP_439893.2
|
4, 5 |
851 |
|
XM_011514221.1 |
XP_011512523.1
|
6, 7 |
1099 |
|
XM_011514222.2 |
XP_011512524.1
|
6, 7 |
1333 |
|
XM_011514223.2 |
XP_011512525.1
|
7, 8 |
1214 |
|
XM_011514225.1 |
XP_011512527.1
|
6, 7 |
1099 |
|
GenBank mRNA |
AF220122.1 |
- |
4, 5 |
849 |
|
AF220123.1 |
- |
4, 5 |
849 |
|
AL360163.1 |
- |
2, 3 |
114 |
|
AL360164.1 |
- |
2, 3 |
145 |
|
BC093926.1 |
- |
4, 5 |
829 |
7939771 |
BC113419.1 |
- |
4, 5 |
829 |
8322471 |
View Full Product Details