Gene Transcripts

Gene Symbol
LPIN2
Entrez Gene ID
9663
Gene Name
lipin 2
Gene Aliases
Chromosome Location
Chr. 18: 2916994 - 3013315 on Build GRCh38
UniGene ID
Hs.132342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014646.2 NP_055461.1 6 962
XM_005258177.4 XP_005258234.1 6 1032
XM_005258178.3 XP_005258235.1 6 961
XM_005258179.4 XP_005258236.1 6 895
XM_017026098.1 XP_016881587.1 7 1051
XM_017026099.1 XP_016881588.1 6 822
GenBank mRNA BC152448.1 - 6 962
D87436.1 - 6 962

View Full Product Details