Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014740.3 | NP_055555.1 | 7 | 827 | |
XM_011525522.1 | XP_011523824.1 | 6 | 720 | ||
GenBank mRNA | AK290608.1 | - | 7 | 808 | |
BC003662.2 | - | 7 | 737 | 2988931 | |
BC004386.1 | - | 7 | 758 | 3619386 | |
BC011151.1 | - | 7 | 762 | 3452439 | |
CR456750.1 | - | 7 | 605 | ||
CR749455.1 | - | 1 | 344 | ||
D21853.1 | - | 7 | 819 | ||
X79538.1 | - | 7 | 678 |