Gene Transcripts
- Gene Symbol
- SRPX
- Entrez Gene ID
- 8406
- Gene Name
- sushi repeat containing protein, X-linked
- Gene Aliases
- DRS, ETX1, HEL-S-83p, SRPX1
- Chromosome Location
- Chr. X: 38149335 - 38220924 on Build GRCh38
- UniGene ID
- Hs.15154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170751.1 |
NP_001164222.1
|
2, 3 |
284 |
|
NM_001170752.1 |
NP_001164223.1
|
2, 3 |
284 |
|
NM_006307.4 |
NP_006298.1
|
2, 3 |
284 |
|
XM_017029893.1 |
XP_016885382.1
|
2, 3 |
223 |
|
GenBank mRNA |
AK289430.1 |
- |
2, 3 |
231 |
|
AK293127.1 |
- |
2, 3 |
261 |
|
AK298802.1 |
- |
2, 3 |
220 |
|
BC020684.1 |
- |
2, 3 |
239 |
4731946 |
DC388035.1 |
- |
2, 3 |
284 |
|
EU668359.1 |
- |
2, 3 |
190 |
|
U61374.1 |
- |
2, 3 |
190 |
|
U78093.1 |
- |
2, 3 |
191 |
|
View Full Product Details