Gene Transcripts

Gene Symbol
RAB11FIP3
Entrez Gene ID
9727
Gene Name
RAB11 family interacting protein 3
Gene Aliases
CART1, Rab11-FIP3
Chromosome Location
Chr. 16: 425621 - 522485 on Build GRCh38
UniGene ID
Hs.531642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142272.1 NP_001135744.1 12 1889
NM_014700.3 NP_055515.1 14 2935
XM_005255713.3 XP_005255770.1 15 3105
XM_005255714.1 XP_005255771.1 13 1950
XM_005255715.4 XP_005255772.1 13 2756
XM_011522764.2 XP_011521066.1 13 1982
XM_011522765.2 XP_011521067.1 13 1808
GenBank mRNA AB014565.1 - 14 2983
AF038180.1 - 1 380
AF395731.1 - 14 2983
AY130007.1 - 6 1004
BC051360.1 - 14 2934 6646253

View Full Product Details