Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304777.1 | NP_001291706.1 | 5 | 440 | |
NM_016041.4 | NP_057125.2 | 5 | 443 | ||
NR_130905.1 | 4 | 555 | |||
NR_130906.1 | 4 | 349 | |||
GenBank mRNA | AF132289.1 | - | 5 | 391 | |
AF208065.1 | - | 5 | 379 | ||
AF242523.1 | - | 5 | 379 | ||
AK024574.1 | - | 1 | 61 | ||
BC010890.1 | - | 5 | 391 | 4280273 | |
BG110482.1 | - | 4 | 504 | ||
CN362881.1 | - | 5 | 361 | ||
CR457202.1 | - | 5 | 374 | ||
HY051246.1 | - | 3, 4 | 309 |