Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr. 17: 38800434 - 38825350 on Build GRCh38
UniGene ID
Hs.406223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017748.4 NP_060218.1 5 753
NR_073428.1 6 870
GenBank mRNA AK000298.1 - 5 703
AK225373.1 - 5 723
AK296115.1 - 6 835
AK302983.1 - 5 654
AK316077.1 - 4 507
BC008833.2 - 5 739 4304656

View Full Product Details