Gene Transcripts

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
Chromosome Location
Chr. 15: 40735884 - 40755336 on Build GRCh38
UniGene ID
Hs.511067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304802.1 NP_001291731.1 12, 13 1598
NM_001323894.1 NP_001310823.1 12, 13 1512
NM_001323895.1 NP_001310824.1 12, 13 1729
NM_001323896.1 NP_001310825.1 12, 13 1590
NM_001323897.1 NP_001310826.1 12, 13 1706
NM_018145.2 NP_060615.1 12, 13 1628
XM_011521755.2 XP_011520057.1 9, 10 1874
GenBank mRNA AB000782.1 - 11, 12 1367
AK001441.1 - 12, 13 1552
AK090731.1 - 5, 6 1500
AK092058.1 - 12, 13 1511
AK097286.1 - 13, 14 1440
AK123192.1 - 13, 14 1505
AK123282.1 - 11, 12 2435
AY358793.1 - 11, 12 1368
BC008970.2 - 8, 9 825
BC063844.1 - 12, 13 1516 6527521

View Full Product Details