Gene Transcripts

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
Chromosome Location
Chr. 17: 3806748 - 3846251 on Build GRCh38
UniGene ID
Hs.120963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114118.2 NP_001107590.1 12 1512
XM_005256717.2 XP_005256774.1 12 1512
XM_006721556.2 XP_006721619.1 11 1401
XM_011523954.2 XP_011522256.1 12 1429
XM_011523955.2 XP_011522257.1 11 1283
XM_017024826.1 XP_016880315.1 11 1399
XM_017024827.1 XP_016880316.1 6 986
XM_017024828.1 XP_016880317.1 7 804
XM_017024829.1 XP_016880318.1 7 804
GenBank mRNA AJ277841.1 - 7 809
AK125048.1 - 9 2201
AK223470.1 - 5 688
BC010707.1 - 5 627

View Full Product Details