Gene Transcripts

Gene Symbol
RPAP3
Entrez Gene ID
79657
Gene Name
RNA polymerase II associated protein 3
Gene Aliases
Chromosome Location
Chr. 12: 47661932 - 47706061 on Build GRCh38
UniGene ID
Hs.437855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146075.1 NP_001139547.1 9 1025
NM_001146076.1 NP_001139548.1 8 866
NM_024604.2 NP_078880.2 9 1025
XM_011538741.2 XP_011537043.1 7 776
GenBank mRNA AK025561.1 - 9 955
AK299465.1 - 8 831
BC056415.1 - 9 950 5218249

View Full Product Details