Gene Transcripts

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
Chromosome Location
Chr. 15: 67200675 - 67255198 on Build GRCh38
UniGene ID
Hs.254642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271885.1 NP_001258814.1 3 307
NM_001271886.1 NP_001258815.1 3 568
NM_024666.4 NP_078942.3 3 423
XM_011522020.1 XP_011520322.1 3 673
GenBank mRNA AK021568.1 - 3 327
AK294402.1 - 3 304
AL136715.1 - 3 333
BC047026.1 - 3 346 5534878
BC058886.1 - 3 345 6503238
CR533540.1 - 3 318
DA688370.1 - 3 423
DC315715.1 - 3 307

View Full Product Details