Gene Transcripts

Gene Symbol
KDM8
Entrez Gene ID
79831
Gene Name
lysine demethylase 8
Gene Aliases
JMJD5
Chromosome Location
Chr. 16: 27203477 - 27222305 on Build GRCh38
UniGene ID
Hs.145717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145348.1 NP_001138820.1 8 1528
NM_024773.2 NP_079049.2 8 1478
XM_017023676.1 XP_016879165.1 7 1385
XM_017023677.1 XP_016879166.1 9 1563
XM_017023678.1 XP_016879167.1 9 1559
XM_017023679.1 XP_016879168.1 9 1559
XM_017023680.1 XP_016879169.1 6 1070
GenBank mRNA AK023860.1 - 8 1469
AK225066.1 - 8 1435
AK297166.1 - 8 1528
AK298410.1 - 6 518
AK308499.1 - 6 728
BC027911.1 - 8 1457 5207043

View Full Product Details