Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286746.1 | NP_001273675.1 | 10 | 1532 | |
NM_001286747.1 | NP_001273676.1 | 9 | 1327 | ||
NM_024808.3 | NP_079084.3 | 10 | 1434 | ||
XM_006719868.3 | XP_006719931.2 | 8 | 1291 | ||
GenBank mRNA | AK026277.1 | - | 5 | 759 | |
AK291496.1 | - | 10 | 1260 | ||
AK298608.1 | - | 9 | 1153 | ||
BC007119.1 | - | 1 | 218 | ||
BC025367.2 | - | 10 | 1245 | 4098541 | |
BC044213.1 | - | 9 | 1058 | ||
BC056876.1 | - | 10 | 1257 | 6473892 | |
BX648605.1 | - | 10 | 1532 | ||
EU834129.1 | - | 10 | 1340 |