Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
80301
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
PLEKHQ1, PP1628, pp9099
Chromosome Location
Chr. 15: 64841883 - 64868002 on Build GRCh38
UniGene ID
Hs.709337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195059.1 NP_001181988.1 2 229
NM_025201.4 NP_079477.2 3 379
GenBank mRNA AF494534.1 - 3 328
AK096203.1 - 3 347
BC008744.2 - 2 190 3346334

View Full Product Details