Gene Transcripts

Gene Symbol
INF2
Entrez Gene ID
64423
Gene Name
inverted formin, FH2 and WH2 domain containing
Gene Aliases
C14orf151, C14orf173, CMTDIE, FSGS5, pp9484
Chromosome Location
Chr. 14: 104689606 - 104719610 on Build GRCh38
UniGene ID
Hs.24956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032714.2 NP_116103.1 5 1017
GenBank mRNA AK290083.1 - 5 1002
BC006173.1 - 5 1002

View Full Product Details