Gene Transcripts
- Gene Symbol
- LRRC45
- Entrez Gene ID
- 201255
- Gene Name
- leucine rich repeat containing 45
- Gene Aliases
- Chromosome Location
- Chr. 17: 82023252 - 82031151 on Build GRCh38
- UniGene ID
- Hs.143774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144999.3 |
NP_659436.1
|
10, 11 |
1472 |
|
GenBank mRNA |
BC014109.2 |
- |
10, 11 |
1370 |
|
View Full Product Details