Gene Transcripts

Gene Symbol
TVP23C
Entrez Gene ID
201158
Gene Name
trans-golgi network vesicle protein 23 homolog C (S. cerevisiae)
Gene Aliases
FAM18B2
Chromosome Location
Chr. 17: 15502264 - 15563631 on Build GRCh38
UniGene ID
Hs.164595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145301.2 NP_660344.2 6 817
GenBank mRNA BC011952.1 - 6 717

View Full Product Details