Gene Transcripts
- Gene Symbol
- CPNE8
- Entrez Gene ID
- 144402
- Gene Name
- copine 8
- Gene Aliases
- Chromosome Location
- Chr. 12: 38652200 - 38908593 on Build GRCh38
- UniGene ID
- Hs.40910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153634.2 |
NP_705898.1
|
6, 7 |
492 |
|
XM_011537951.2 |
XP_011536253.1
|
6, 7 |
3467 |
|
XM_011537952.2 |
XP_011536254.1
|
6, 7 |
3467 |
|
XM_017018851.1 |
XP_016874340.1
|
7, 8 |
1465 |
|
XM_017018852.1 |
XP_016874341.1
|
6, 7 |
438 |
|
GenBank mRNA |
AK098593.1 |
- |
6, 7 |
422 |
|
AK307908.1 |
- |
6, 7 |
465 |
|
AY177785.1 |
- |
6, 7 |
408 |
|
BC068040.1 |
- |
6, 7 |
456 |
|
BC110577.1 |
- |
6, 7 |
483 |
40007743 |
BC110578.1 |
- |
6, 7 |
483 |
40007744 |
BI669075.1 |
- |
6, 7 |
465 |
|
BX538055.1 |
- |
6, 7 |
506 |
|
View Full Product Details