Gene Transcripts

Gene Symbol
CFAP161
Entrez Gene ID
161502
Gene Name
cilia and flagella associated protein 161
Gene Aliases
C15orf26
Chromosome Location
Chr. 15: 81099408 - 81149175 on Build GRCh38
UniGene ID
Hs.130979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173528.2 NP_775799.2 2, 3 192
XM_006720408.2 XP_006720471.1 3, 4 345
XM_011521280.1 XP_011519582.1 2, 3 186
XM_017021963.1 XP_016877452.1 3, 4 270
GenBank mRNA AK095934.1 - 3, 4 345
AK302120.1 - 2, 3 230
AK307565.1 - 2, 3 187
BC062471.1 - 2, 3 192 30390735

View Full Product Details