Gene Transcripts

Gene Symbol
MAPK13
Entrez Gene ID
5603
Gene Name
mitogen-activated protein kinase 13
Gene Aliases
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta
Chromosome Location
Chr. 6: 36130484 - 36144524 on Build GRCh38
UniGene ID
Hs.178695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002754.4 NP_002745.1 9, 10 867
GenBank mRNA AB451221.1 - 10, 11 768
AB451344.1 - 10, 11 768
AF004709.1 - 9, 10 816
AF015256.1 - 9, 10 828
AF092535.1 - 9, 10 789
AF100546.1 - 9, 10 770
AK308018.1 - 9, 10 848
BC000433.2 - 9, 10 809 2819932
BC001641.2 - 9, 10 809 2819932
BC004428.1 - 9, 10 809 2819932
BT007221.1 - 9, 10 768
CR536490.1 - 9, 10 768
CR541687.1 - 9, 10 768
KU178281.1 - 9, 10 768
KU178282.1 - 9, 10 801
U93232.1 - 9, 10 775
Y10488.1 - 9, 10 790

View Full Product Details