Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191037.1 | NP_001177966.1 | 8 | 1081 | |
NM_002807.3 | NP_002798.2 | 8 | 1081 | ||
NR_034059.1 | 7 | 911 | |||
XM_017004517.1 | XP_016860006.1 | 8 | 1081 | ||
GenBank mRNA | AK307415.1 | - | 7 | 908 | |
AK307513.1 | - | 6 | 783 | ||
AK312528.1 | - | 8 | 1038 | ||
BC001053.2 | - | 8 | 1038 | ||
BC014013.2 | - | 8 | 1050 | ||
BC020223.1 | - | 8 | 1028 | ||
BC032445.1 | - | 8 | 1053 | ||
BC039845.1 | - | 8 | 1004 | ||
BC047897.1 | - | 8 | 1003 | ||
BC064398.1 | - | 8 | 1052 | ||
BC073833.1 | - | 8 | 1005 | ||
BC094720.1 | - | 8 | 1067 | 4826340 | |
BC112344.1 | - | 8 | 1042 | 40008454 | |
BC114434.1 | - | 8 | 1042 | 40008444 | |
D44466.1 | - | 8 | 999 | ||
KU921434.1 | - | 1 | 38 |