Gene Transcripts

Gene Symbol
ATIC
Entrez Gene ID
471
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Gene Aliases
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH
Chromosome Location
Chr. 2: 215311956 - 215359745 on Build GRCh38
UniGene ID
Hs.90280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004044.6 NP_004035.2 14, 15 1698
XM_017004187.1 XP_016859676.1 14, 15 1621
GenBank mRNA AB062403.1 - 14, 15 1546
AK290067.1 - 13, 14 1680
AK298142.1 - 14, 15 1811
AK313066.1 - 14, 15 1592
BC008879.2 - 14, 15 1541 4300570
D82348.1 - 14, 15 1586
D89976.1 - 14, 15 1568
EU794654.1 - 14, 15 1685
U37436.1 - 13, 14 1508

View Full Product Details