Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001247997.1 | NP_001234926.1 | 3 | 476 | |
NM_002956.2 | NP_002947.1 | 3 | 476 | ||
NM_198240.1 | NP_937883.1 | 3 | 476 | ||
XM_005253593.2 | XP_005253650.1 | 3 | 539 | ||
XM_006719552.3 | XP_006719615.1 | 3 | 565 | ||
XM_017019786.1 | XP_016875275.1 | 3 | 640 | ||
XM_017019787.1 | XP_016875276.1 | 3 | 539 | ||
XM_017019788.1 | XP_016875277.1 | 3 | 565 | ||
XM_017019789.1 | XP_016875278.1 | 3 | 724 | ||
XM_017019790.1 | XP_016875279.1 | 3 | 466 | ||
XM_017019791.1 | XP_016875280.1 | 3 | 565 | ||
XM_017019792.1 | XP_016875281.1 | 3 | 565 | ||
GenBank mRNA | BC047895.1 | - | 3 | 482 | |
BC062543.1 | - | 2 | 432 | ||
BC114213.1 | - | 2 | 391 | 7961952 | |
BC117209.1 | - | 2 | 363 | 40125760 | |
BC126305.1 | - | 2 | 363 | 8992021 | |
M97501.1 | - | 3 | 476 | ||
X64838.1 | - | 3 | 453 |