Gene Transcripts

Gene Symbol
SLN
Entrez Gene ID
6588
Gene Name
sarcolipin
Gene Aliases
Chromosome Location
Chr. 11: 107707375 - 107712061 on Build GRCh38
UniGene ID
Hs.334629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003063.2 NP_003054.1 2 691
GenBank mRNA BC005261.1 - 2 686
BC094685.1 - 2 673 6203008
F30315.1 - 1 305
U96094.1 - 2 685

View Full Product Details