Gene Transcripts

Gene Symbol
SRSF1
Entrez Gene ID
6426
Gene Name
serine and arginine rich splicing factor 1
Gene Aliases
ASF, SF2, SF2p33, SFRS1, SRp30a
Chromosome Location
Chr. 17: 58000919 - 58007346 on Build GRCh38
UniGene ID
Hs.68714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078166.1 NP_001071634.1 2 567
NM_006924.4 NP_008855.1 2 567
NR_034041.1 2 567
GenBank mRNA AB062124.1 - 2 448
AB209558.1 - 2 454
AK225711.1 - 2 445
AK289933.1 - 2 476
AK312781.1 - 2 464
BC010264.1 - 2 393 2900101
BC033785.1 - 2 453 5166554
M69040.1 - 2 482
M72709.1 - 2 454

View Full Product Details