Gene Transcripts

Gene Symbol
CFP
Entrez Gene ID
5199
Gene Name
complement factor properdin
Gene Aliases
BFD, PFC, PFD, PROPERDIN
Chromosome Location
Chr. X: 47624213 - 47630305 on Build GRCh38
UniGene ID
Hs.53155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145252.1 NP_001138724.1 5 776
NM_002621.2 NP_002612.1 6 892
XM_017029575.1 XP_016885064.1 4 1398
GenBank mRNA AK122955.1 - 4 1948
AK310695.1 - 5 771
BC015756.1 - 5 710 4850121
KU178249.1 - 5 650
KU178250.1 - 5 650
M83652.1 - 5 722
X57748.1 - 5 715

View Full Product Details