Gene Transcripts
- Gene Symbol
- LMO7
- Entrez Gene ID
- 4008
- Gene Name
- LIM domain 7
- Gene Aliases
- FBX20, FBXO20, LOMP
- Chromosome Location
- Chr. 13: 75620434 - 75859870 on Build GRCh38
- UniGene ID
- Hs.207631
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005358.5 |
NP_005349.3
|
2, 3 |
1477 |
|
GenBank mRNA |
AF330045.1 |
- |
2, 3 |
1476 |
|
View Full Product Details