Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324255.1 | NP_001311184.1 | 2 | 344 | |
NM_005909.4 | NP_005900.2 | 3 | 614 | ||
GenBank mRNA | AK096273.1 | - | 4 | 437 | |
AK307990.1 | - | 2 | 344 | ||
AK308147.1 | - | 3 | 405 | ||
BC017240.2 | - | 3 | 591 | ||
BC025240.1 | - | 3 | 533 | ||
BC032516.1 | - | 3 | 406 | ||
BC039822.1 | - | 3 | 400 | ||
BC046114.1 | - | 3 | 420 | ||
BC056145.1 | - | 3 | 394 | ||
BC063669.1 | - | 3 | 394 | ||
BC073993.1 | - | 3 | 396 | ||
BC094834.1 | - | 3 | 391 | ||
BC108733.1 | - | 3 | 409 | ||
BC139918.1 | - | 3 | 511 | ||
BC141853.1 | - | 2 | 197 | ||
BC150196.1 | - | 3 | 511 | ||
BX647077.1 | - | 3 | 405 |