Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr. 17: 58968068 - 59106921 on Build GRCh38
UniGene ID
Hs.579079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005207.3 NP_001005207.1 6 879
NM_001320987.1 NP_001307916.1 5 777
NM_001320988.1 NP_001307917.1 6 879
NM_001320989.1 NP_001307918.1 6 879
NM_001320990.1 NP_001307919.1 7 955
NM_015294.4 NP_056109.1 6 879
XM_005257385.1 XP_005257442.1 6 711
XM_011524831.1 XP_011523133.1 6 711
XM_011524832.2 XP_011523134.1 6 711
XM_011524833.1 XP_011523135.1 6 711
XM_011524834.1 XP_011523136.1 5 593
XM_011524836.1 XP_011523138.1 6 711
XM_017024662.1 XP_016880151.1 6 711
XM_017024663.1 XP_016880152.1 6 711
XM_017024664.1 XP_016880153.1 6 711
XM_017024665.1 XP_016880154.1 5 593
XM_017024666.1 XP_016880155.1 6 711
XM_017024667.1 XP_016880156.1 6 711
XM_017024668.1 XP_016880157.1 5 593
XM_017024669.1 XP_016880158.1 5 593
XM_017024670.1 XP_016880159.1 5 592
XM_017024671.1 XP_016880160.1 5 623
XM_017024672.1 XP_016880161.1 5 623
XM_017024673.1 XP_016880162.1 5 398
XM_017024674.1 XP_016880163.1 5 398
GenBank mRNA AB020705.1 - 6 464
AF213365.1 - 6 683
AK289674.1 - 6 694
AK292459.1 - 6 751
AK294850.1 - 5 682
BC036012.1 - 6 715 5270430
BX537955.1 - 7 774

View Full Product Details