Gene Transcripts
- Gene Symbol
- IL12A
- Entrez Gene ID
- 3592
- Gene Name
- interleukin 12A
- Gene Aliases
- CLMF, IL-12A, NFSK, NKSF1, P35
- Chromosome Location
- Chr. 3: 159988836 - 159996019 on Build GRCh38
- UniGene ID
- Hs.673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000882.3 |
NP_000873.2
|
7 |
1341 |
|
GenBank mRNA |
M65291.1 |
- |
7 |
1221 |
|
View Full Product Details