Gene Transcripts
- Gene Symbol
- GBF1
- Entrez Gene ID
- 8729
- Gene Name
- golgi brefeldin A resistant guanine nucleotide exchange factor 1
- Gene Aliases
- ARF1GEF
- Chromosome Location
- Chr. 10: 102245498 - 102382899 on Build GRCh38
- UniGene ID
- Hs.290243
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199378.1 |
NP_001186307.1
|
24, 25 |
3440 |
|
NM_001199379.1 |
NP_001186308.1
|
24, 25 |
3437 |
|
NM_004193.2 |
NP_004184.1
|
24, 25 |
3437 |
|
XM_005270261.1 |
XP_005270318.1
|
24, 25 |
3440 |
|
XM_006718047.1 |
XP_006718110.1
|
25, 26 |
3539 |
|
XM_006718048.1 |
XP_006718111.1
|
25, 26 |
3536 |
|
XM_006718049.1 |
XP_006718112.1
|
25, 26 |
3539 |
|
XM_006718050.1 |
XP_006718113.1
|
25, 26 |
3503 |
|
XM_011540312.1 |
XP_011538614.1
|
24, 25 |
3398 |
|
XM_011540313.1 |
XP_011538615.1
|
25, 26 |
3539 |
|
XM_011540314.2 |
XP_011538616.1
|
13, 14 |
1917 |
|
XM_017016861.1 |
XP_016872350.1
|
25, 26 |
3515 |
|
XM_017016862.1 |
XP_016872351.1
|
25, 26 |
3503 |
|
XM_017016863.1 |
XP_016872352.1
|
24, 25 |
3401 |
|
XM_017016864.1 |
XP_016872353.1
|
20, 21 |
2739 |
|
GenBank mRNA |
AF068755.1 |
- |
24, 25 |
3383 |
|
AK024916.1 |
- |
4, 5 |
550 |
|
AK025330.1 |
- |
24, 25 |
3406 |
|
BC117681.1 |
- |
24, 25 |
3264 |
40070852 |
BC117682.1 |
- |
24, 25 |
3264 |
40070853 |
D87435.2 |
- |
24, 25 |
3367 |
|
View Full Product Details