Gene Transcripts
- Gene Symbol
- ILF3
- Entrez Gene ID
- 3609
- Gene Name
- interleukin enhancer binding factor 3
- Gene Aliases
- CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80
- Chromosome Location
- Chr. 19: 10654261 - 10692419 on Build GRCh38
- UniGene ID
- Hs.465885
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001137673.1 |
NP_001131145.1
|
12, 13 |
1705 |
|
NM_004516.3 |
NP_004507.2
|
12, 13 |
1705 |
|
NM_012218.3 |
NP_036350.2
|
12, 13 |
1705 |
|
NM_017620.2 |
NP_060090.2
|
12, 13 |
1705 |
|
NM_153464.2 |
NP_703194.1
|
12, 13 |
1705 |
|
XM_006722742.2 |
XP_006722805.1
|
12, 13 |
1620 |
|
XM_011527984.2 |
XP_011526286.2
|
12, 13 |
1738 |
|
XM_011527985.2 |
XP_011526287.1
|
12, 13 |
1620 |
|
XM_011527987.1 |
XP_011526289.1
|
8, 9 |
999 |
|
XM_017026763.1 |
XP_016882252.1
|
13, 14 |
1829 |
|
XM_017026764.1 |
XP_016882253.1
|
9, 10 |
1293 |
|
XM_017026765.1 |
XP_016882254.1
|
11, 12 |
1522 |
|
GenBank mRNA |
AB209062.1 |
- |
11, 12 |
2325 |
|
AF141870.1 |
- |
12, 13 |
1499 |
|
AF147209.1 |
- |
10, 11 |
1400 |
|
AF167569.1 |
- |
10, 11 |
1388 |
|
AF167570.1 |
- |
10, 11 |
1388 |
|
AJ271744.1 |
- |
8, 9 |
988 |
|
AJ271745.1 |
- |
11, 12 |
1477 |
|
AJ271746.1 |
- |
11, 12 |
1457 |
|
AK291205.1 |
- |
12, 13 |
1604 |
|
AK291617.1 |
- |
12, 13 |
1590 |
|
AK293170.1 |
- |
4, 5 |
437 |
|
AK294084.1 |
- |
8, 9 |
1008 |
|
BC064836.1 |
- |
12, 13 |
1638 |
6503234 |
HQ322263.1 |
- |
10, 11 |
1388 |
|
HQ322264.1 |
- |
10, 11 |
1388 |
|
HQ322265.1 |
- |
10, 11 |
1388 |
|
U10324.1 |
- |
12, 13 |
1655 |
|
X98265.1 |
- |
11, 12 |
1597 |
|
View Full Product Details