Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001967.3 | NP_001958.2 | 3 | 227 | |
GenBank mRNA | AB209021.1 | - | 2 | 161 | |
AF208852.1 | - | 3 | 205 | ||
AK130087.1 | - | 3 | 222 | ||
AK225646.1 | - | 3 | 222 | ||
AL117412.1 | - | 3 | 227 | ||
AL556981.3 | - | 3 | 227 | ||
AY207392.1 | - | 2 | 158 | ||
BC012547.1 | - | 3 | 229 | 4079371 | |
BC013708.1 | - | 3 | 207 | 4341420 | |
BC015842.1 | - | 3 | 226 | 4337581 | |
BC048105.1 | - | 3 | 225 | 4820774 | |
BT009860.1 | - | 3 | 188 | ||
D30655.1 | - | 3 | 203 | ||
S79942.1 | - | 3 | 203 |