Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
1411
Gene Name
crystallin beta A1
Gene Aliases
CRYB1, CTRCT10
Chromosome Location
Chr. 17: 29246857 - 29254494 on Build GRCh38
UniGene ID
Hs.46275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005208.4 NP_005199.2 6 674
XM_017024198.1 XP_016879687.1 7 970
GenBank mRNA BC069537.1 - 6 667
BC113693.1 - 6 668 8322745
KM230602.1 - 6 656
KM230603.1 - 6 655
KM230604.1 - 6 652
KM230605.1 - 5 652
KR336660.1 - 6 652
KR336661.1 - 3 341

View Full Product Details