Gene Transcripts

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
Chromosome Location
Chr. 7: 143390813 - 143408892 on Build GRCh38
UniGene ID
Hs.89839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005232.4 NP_005223.4 12, 13 2177
XM_006715880.3 XP_006715943.1 11, 12 1805
GenBank mRNA AK313263.1 - 12, 13 2150
BC130291.1 - 12, 13 2161 40146322
BC144156.1 - 11, 12 2313
CN289901.1 - 5, 6 538
M18391.1 - 12, 13 2207
Z27409.1 - 8, 9 1235

View Full Product Details