Gene Transcripts

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
Chromosome Location
Chr. X: 53431258 - 53434376 on Build GRCh38
UniGene ID
Hs.171280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037811.2 NP_001032900.1 2 183
NM_004493.2 NP_004484.1 2 183
GenBank mRNA AF035555.2 - 2 188
AF069134.1 - 2 170
AU132847.1 - 2 183
BC000372.2 - 2 167 2819721
BC000829.1 - 2 167
BC008708.1 - 2 163 3160844
CR456723.1 - 2 152
KU178107.1 - 2 152
KU178108.1 - 2 152
U73514.1 - 2 167
U96132.1 - 2 179

View Full Product Details