Gene Transcripts
- Gene Symbol
- DDX11
- Entrez Gene ID
- 1663
- Gene Name
- DEAD/H-box helicase 11
- Gene Aliases
- CHL1, CHLR1, KRG2, WABS
- Chromosome Location
- Chr. 12: 31073574 - 31104799 on Build GRCh38
- UniGene ID
- Hs.443960
- Species
- Homo sapiens
- Gene Symbol
- DDX12P
- Entrez Gene ID
- 440081
- Gene Name
- DEAD/H-box helicase 12, pseudogene
- Gene Aliases
- CHLR2, DDX12
- Chromosome Location
- Chr. 12: 9417691 - 9448172 on Build GRCh38
- UniGene ID
- Hs.447869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033399.1 |
|
27 |
3823 |
|
GenBank mRNA |
BC065494.1 |
- |
15 |
3837 |
|
DQ786301.1 |
- |
1 |
730 |
|
- Gene Symbol
- LOC642846
- Entrez Gene ID
- 642846
- Gene Name
- DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11-like
- Gene Aliases
- Chromosome Location
- Chr. 12: 9283657 - 9314088 on Build GRCh38
- UniGene ID
- Hs.447869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024374.1 |
|
22 |
3435 |
|
View Full Product Details