Gene Transcripts
- Gene Symbol
- CASP10
- Entrez Gene ID
- 843
- Gene Name
- caspase 10
- Gene Aliases
- ALPS2, FLICE2, MCH4
- Chromosome Location
- Chr. 2: 201182881 - 201229406 on Build GRCh38
- UniGene ID
- Hs.5353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206542.1 |
NP_001193471.1
|
6, 7 |
1205 |
|
NM_001230.4 |
NP_001221.2
|
6, 7 |
1205 |
|
NM_032974.4 |
NP_116756.2
|
8, 9 |
1334 |
|
NM_032976.3 |
NP_116758.1
|
7, 8 |
1242 |
|
NM_032977.3 |
NP_116759.2
|
8, 9 |
1334 |
|
XM_005246907.2 |
XP_005246964.1
|
8, 9 |
1117 |
|
XM_006712796.3 |
XP_006712859.1
|
7, 8 |
1252 |
|
XM_011511990.2 |
XP_011510292.1
|
7, 8 |
1025 |
|
GenBank mRNA |
AF111344.1 |
- |
6, 7 |
824 |
|
AF111345.1 |
- |
7, 8 |
916 |
|
AJ487679.1 |
- |
6, 7 |
908 |
|
BC042844.1 |
- |
8, 9 |
1070 |
5176698 |
KU177959.1 |
- |
7, 8 |
916 |
|
KU177960.1 |
- |
5, 6 |
787 |
|
U60519.1 |
- |
6, 7 |
934 |
|
U86214.1 |
- |
8, 9 |
1110 |
|
View Full Product Details