Gene Transcripts

Gene Symbol
PHC2
Entrez Gene ID
1912
Gene Name
polyhomeotic homolog 2
Gene Aliases
EDR2, HPH2, PH2
Chromosome Location
Chr. 1: 33323623 - 33431017 on Build GRCh38
UniGene ID
Hs.524271
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004427.3 NP_004418.2 7 1671
NM_198040.2 NP_932157.1 14 2978
XM_005270570.1 XP_005270627.1 7 1607
XM_017000513.1 XP_016856002.1 15 3477
XM_017000514.1 XP_016856003.1 15 3089
XM_017000515.1 XP_016856004.1 15 3182
XM_017000516.1 XP_016856005.1 15 3411
XM_017000517.1 XP_016856006.1 15 3023
XM_017000518.1 XP_016856007.1 15 3116
XM_017000519.1 XP_016856008.1 16 3566
XM_017000520.1 XP_016856009.1 14 2936
XM_017000521.1 XP_016856010.1 14 3355
GenBank mRNA AJ419231.1 - 14 2978
AK056429.1 - 8 1908
AK091856.1 - 1 1407
AK128821.1 - 8 1656
BC015450.1 - 3 818
BC018602.1 - 7 1677
BC028396.1 - 7 1642
BC029269.1 - 7 1513
BC068573.1 - 7 1565
BC092492.1 - 7 1565
BC110863.1 - 7 1564
BC130630.1 - 13 2894 40146661
BC144121.1 - 13 2894 9052641
BX537498.1 - 1 1643

View Full Product Details