Gene Transcripts

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
Chromosome Location
Chr. 20: 38304150 - 38337503 on Build GRCh38
UniGene ID
Hs.529019
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001725.2 NP_001716.2 4 491
GenBank mRNA AF322588.1 - 4 417
AK296568.1 - 4 336
AK315328.1 - 4 458
AX713278.1 - 4 459
AX746123.1 - 4 459
AX752831.1 - 4 459
BC040955.1 - 4 466 5218655
BP306215.1 - 4 503
DQ414688.1 - 4 429
J04739.1 - 4 459

View Full Product Details