Gene Transcripts

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
Chromosome Location
Chr. 11: 61752617 - 61788518 on Build GRCh38
UniGene ID
Hs.473109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127392.2 NP_001120864.1 10 1570
NM_013279.3 NP_037411.1 10 1470
XM_005274222.1 XP_005274279.1 10 1558
XM_005274223.1 XP_005274280.1 10 1558
XM_005274224.1 XP_005274281.1 10 1558
XM_005274225.1 XP_005274282.1 10 1558
XM_005274226.1 XP_005274283.1 10 1558
XM_005274227.1 XP_005274284.1 10 1558
XM_005274228.1 XP_005274285.1 10 1405
XM_011545234.2 XP_011543536.1 10 1444
GenBank mRNA AB023171.2 - 10 1558
AF038536.1 - 38
AF086762.1 - 10 1470

View Full Product Details