Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317199.1 | NP_001304128.1 | 4 | 1848 | |
NM_001317200.1 | NP_001304129.1 | 3 | 1285 | ||
NM_001317201.1 | NP_001304130.1 | 3 | 1337 | ||
NM_001317202.1 | NP_001304131.1 | 3 | 1301 | ||
NM_001317203.1 | NP_001304132.1 | 4 | 1509 | ||
NM_001317204.1 | NP_001304133.1 | 4 | 1379 | ||
NM_001317205.1 | NP_001304134.1 | 3 | 1378 | ||
NM_001317206.1 | NP_001304135.1 | 5 | 1605 | ||
NM_001317207.1 | NP_001304136.1 | 3 | 1287 | ||
NM_001317208.1 | NP_001304137.1 | 4 | 1393 | ||
NM_001317209.1 | NP_001304138.1 | 3 | 1299 | ||
NM_001317210.1 | NP_001304139.1 | 4 | 1387 | ||
NM_001317211.1 | NP_001304140.1 | 4 | 1381 | ||
NM_016463.8 | NP_057547.5 | 3 | 1754 | ||
XM_017009573.1 | XP_016865062.1 | 3 | 1624 | ||
GenBank mRNA | AB097005.1 | - | 3 | 1331 | |
AB097032.1 | - | 3 | 1331 | ||
AF151029.1 | - | 2 | 960 | ||
AK001782.1 | - | 3 | 1276 | ||
AK024338.1 | - | 3 | 1754 | ||
AY007103.1 | - | 2 | 1399 | ||
BC002490.2 | - | 2 | 1090 | 3050465 | |
BC006428.1 | - | 3 | 1685 | 3343683 | |
BC017439.1 | - | 3 | 1646 | 2989328 | |
BC024040.1 | - | 3 | 1675 | 3925666 | |
BC041013.1 | - | 3 | 1675 | 4139961 | |
BC050046.1 | - | 3 | 1647 |